missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)aniline-2,4,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Aminobenzotrifluoride ring-d4 |
| CAS | 1219802-14-8 |
| Molecular Formula | C7 2H4 H2 F3 N |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C7H6F3N/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4 H,11H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(N)c([2 H])c(c1[2 H])C(F)(F)F |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 165.15 |
| Formula Weight | 165.0703 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction