missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butyldimethylsilyloxyoleyl Carnitine 4-Methoxybenzyl Ester, TRC

Product Code. 30336005
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336005 10 mg 10mg
30340190 5 mg 5mg
30345254 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30336005 Supplier Toronto Research Chemicals Supplier No. TRCB41571010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-tert-Butyldimethylsilyloxyoleyl Carnitine 4-Methoxybenzyl Ester
Synonym 3-tert-Butyldimethylsilyloxyoleyl Carnitine 4-Methoxybenzyl Ester∣3-tert-Butyldimethylsilyloxyoleyl Carnitine 4-Methoxybenzyl Ester∣
Quantity 10 mg
Shipping Condition Room Temperature
Molecular Formula C38H68ClNO5Si
Formula Weight 681.46
Molecular Weight (g/mol) 682.49
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.