missing translation for 'onlineSavingsMsg'
Learn More

3-(S)-(4-(S)-Isopropyl-2-oxo-oxazolidine-3-carbonyl)-octanoic Acid tert-Butyl Ester, TRC

Product Code. 30391676
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391676 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391676 Supplier Toronto Research Chemicals Supplier No. TRCI87285050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(S)-(4-(S)-Isopropyl-2-oxo-oxazolidine-3-carbonyl)-octanoi
CAS 147961-55-5
Molecular Formula C19 H33 N O5
Quantity 50 mg
InChI Formula InChI=1S/C19H33NO5/c1-7-8-9-10-14(11-16(21)25-19(4,5)6)17(22)20-15(13(2)3)12-24-18(20)23/h13-15H,7-12H2,1-6H3/t14-,15+/m0/s1
SMILES CCCCC[C@@H](CC(=O)OC(C)(C)C)C(=O)N1[C@H](COC1=O)C(C)C
IUPAC Name tert-butyl (3S)-3-[(4S)-2-oxo-4-propan-2-yl-1,3-oxazolidine-3-carbonyl]octanoate
Molecular Weight (g/mol) 355.47
Formula Weight 355.2359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.