Learn More
Fisherbrand™ Three-Way Swivel Scaffolding Connectors
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Outer:
0 to 13 mm
Unit Size:
Each
Description
- With thumbscrews
- For rods and tubes of 6 to 13mm diameters
Specifications
Specifications
| Diameter (Metric) Outer | 0 to 13 mm |
| Product Type | 3 Rods Bossehead 0-13mm |