Learn More
3-Pyridinealdoxime, 98%, ACROS Organics™
Product Code. 10068541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10068541 5 g Glass bottle 5g
10295370 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10068541 Supplier Thermo Scientific Acros Supplier No. 131800050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 1193-92-6
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006409,MFCD00006409
InChI Key YBKOPFQCLSPTPV-VMPITWQZSA-N
Synonym nicotinaldoxime, 3-pyridinealdoxime, nicotinaldehyde oxime, pyridine-3-aldoxime, 3-pyridinecarboxaldehyde, oxime, nicotinaldehyde, oxime, 3-pyridine-aldoxime, pyridine, 3-formyl-, oxime, hydroxyimino-3-pyridylmethane, pyridinaldoxim
PubChem CID 5371219
IUPAC Name (E)-N-[(pyridin-3-yl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=CN=C1
Specifications
CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 122.13
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Pyridinealdoxime, 98%