Learn More
Fisherbrand™ 3-Ply Sterile Facemasks
Product Code. 17397289 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
17 cm
23 cm
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
17397289 17 cm Pack of 200
17307299 23 cm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 17397289
Estimated Shipment: 18-03-2027
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
3-Ply Sterile Facemasks constructed of polyethylene apertured film and PTFE (polytetrafluoroethylene).
  • Constructed from a combination of pleated clear, polyethylene apertured film and PTFE (polytetrafluoroethylene), with soft tie strings
  • Manufactured in systems complying ISO 13485:2016 requirements
  • Certified Sterile
Specifications
Product Type Cleanroom Face Mask
Color White
Sterility Sterile
Style Pleated with Ties
Certifications/Compliance The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013.
Disposable Single-use
For Use With (Application) Industrial Use Only
Length (Metric) 17 cm