Learn More
3-Phosphonobenzoic Acid, TRC
Product Code. 30411199
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411199 2.5 mg 2.5mg
30414371 5 mg -
30411428 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30411199 Supplier Toronto Research Chemicals Supplier No. TRCP3538052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14899-31-1
Molecular Formula C7H7O5P
Molecular Weight (g/mol) 202.1
IUPAC Name 3-phosphonobenzoic acid
SMILES OC(=O)c1cccc(c1)P(=O)(O)O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C7H7O5P/c8-7(9)5-2-1-3-6(4-5)13(10,11)12/h1-4H,(H,8,9)(H2,10,11,12)
Formula Weight 202.0031
Chemical Name or Material 3-Phosphonobenzoic Acid