Learn More
3-Phenyldihydrofuran-2,5-dione, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 1131-15-3 |
| Molecular Formula | C10H8O3 |
| MDL Number | MFCD00192395 |
| Quantity | 25 g |
| SMILES | O=C(O1)C(C2=CC=CC=C2)CC1=O |
| Molecular Weight (g/mol) | 176.17 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-Phenyldihydrofuran-2,5-dione |