Learn More
3-Phenoxyaniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3586-12-7 |
| Molecular Formula | C12H11NO |
| MDL Number | MFCD00041891 |
| Quantity | 1 g |
| SMILES | NC1=CC=CC(OC2=CC=CC=C2)=C1 |
| Molecular Weight (g/mol) | 185.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Phenoxyaniline |