Learn More
3-Penten-2-one, TRC
Product Code. 30179345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179345 1 g 1g
30179590 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179345 Supplier Toronto Research Chemicals Supplier No. TRCP2377901G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 625-33-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
Synonym 2-Oxo-3-pentene, 2-Penten-4-one, Ethylideneacetone, Methyl 1-Propenyl Ketone, Methyl Propenyl Ketone, NSC 61468
SMILES CC(/C=C/C)=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C5H8O/c1-3-4-5(2)6/h3-4H,1-2H3/b4-3+
Chemical Name or Material 3-Penten-2-one