Learn More
3-(Pentafluorothio)aniline, 97%
Product Code. 15487477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487477 5 g 5g
15477477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15487477 Supplier Thermo Scientific Alfa Aesar Supplier No. H35190.06
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
CAS: 2993-22-8 | C6H6F5NS | 219.17 g/mol
Reduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reduction of pentafluoroaniline (PFA) leads to rapid fluoride elimination to form the aminotetrafluorophenyl radical. Homolytic reactions of perfluoroaromatic compounds(Formation of pentafluorophenyl radicals from pentafluoroaniline, and their reactions with aromatic compounds).
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store in a refrigerator. Store under inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2993-22-8
Molecular Formula C6H6F5NS
Molecular Weight (g/mol) 219.17
MDL Number MFCD00674199
InChI Key NUFLICUHOXHWER-UHFFFAOYSA-N
Synonym 3-aminophenylsulfur pentafluoride, 3-pentafluorothio aniline, 3-pentafluorosulfanyl aniline, 3-aminophenylsulphur pentafluoride, sulfur, 3-aminophenyl pentafluoro-, oc-6-21-9ci, 3-pentafluoro-??-sulfanyl aniline, acmc-1cou4, 3-pentafluorosulfanylaniline, 3-pentafluorosulphanylaniline, 3-pentafluorosulfur aniline
PubChem CID 2779188
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)aniline
SMILES NC1=CC(=CC=C1)S(F)(F)(F)(F)F
Specifications
Quantity 5 g
UN Number UN2811
Solubility Information Soluble in methanol.
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)aniline