missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Methoxybenzoyl)indole, TRC

Product Code. 30475591
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475591 250 mg 250mg
30464785 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475591 Supplier Toronto Research Chemicals Supplier No. TRCM261110250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(p-Methoxybenzoyl)indole
CAS 22051-15-6
Molecular Formula C16H13NO2
Quantity 250 mg
Synonym Indol-3-yl p-Methoxyphenyl Ketone, 1H-Indol-3-yl(4-methoxyphenyl)methanone
InChI Formula InChI=1S/C16H13NO2/c1-19-12-8-6-11(7-9-12)16(18)14-10-17-15-5-3-2-4-13(14)15/h2-10,17H,1H3
SMILES O=C(C1=CNC2=C1C=CC=C2)C3=CC=C(OC)C=C3
Molecular Weight (g/mol) 251.28
Formula Weight 251.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.