Learn More
3' '-O-tert-Butyl-2’-deoxymugineic Acid tert-Butyl Diethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3' '-O-tert-Butyl-2'-deoxymugineic Acid tert-Butyl Diethyl Ester |
| CAS | 958350-41-9 |
| Molecular Formula | C24H44N2O7 |
| Quantity | 5 mg |
| Synonym | (alphaS,2S)-alpha-[[(3S)-3,4-Bis(1,1-dimethylethoxy)-4-oxobutyl]amino]-2-(ethoxycarbonyl)-1-azetidinebutanoic Acid Ethyl Ester |
| SMILES | CCOC(=O)[C@H](CCN1CC[C@H]1C(=O)OCC)NCC[C@H](OC(C)(C)C)C(=O)OC(C)(C)C |
| Molecular Weight (g/mol) | 472.62 |