missing translation for 'onlineSavingsMsg'
Learn More

3-O-Benzyl Estradiol, TRC

Product Code. 30336263
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336263 50 mg 50mg
30345151 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30336263 Supplier Toronto Research Chemicals Supplier No. TRCB27647550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-O-Benzyl Estradiol
CAS 14982-15-1
Molecular Formula C25H30O2
Quantity 50 mg
Synonym (17beta)-3-(Phenylmethoxy)-estra-1,3,5(10)-trien-17-ol, 3-(Benzyloxy)-estra-1,3,5(10)-trien-17beta-ol, BLE 99051, Estradiol 3-Benzyl Ether
InChI Formula InChI=1S/C25H30O2/c1-25-14-13-21-20-10-8-19(27-16-17-5-3-2-4-6-17)15-18(20)7-9-22(21)23(25)11-12-24(25)26/h2-6,8,10,15,21-24,26H,7,9,11-14,16H2,1H3/t21-,22-,23+,24+,25+/m1/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(OCc5ccccc5)ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-3-phenylmethoxy-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-ol
Molecular Weight (g/mol) 362.5
Formula Weight 362.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.