Learn More
3-O-Acetyl-betulinic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-O-Acetyl-betulinic acid |
| CAS | 10376-50-8 |
| Molecular Formula | C32H50O4 |
| Quantity | 25 mg |
| Synonym | (3β)-3-(Acetyloxy)lup-20(29)-en-28-oic Acid, 3aH-Cyclopenta[a]chrysene, lup-20(29)-en-28-oic acid deriv., 3-Acetylbetulinic Acid, 3-O-Acetylbetulinic Acid, 3β-Acetoxy-lup-20(29)-en-28-oic Acid, 3β-Acetoxybetulinic Acid, Acetobetulinic Acid, Acetylbetulinic Acid, Betulic Acid Acetate, Betulinic Acid 3-O-Acetate Betulinic acid acetate MJ 321-RS Mairin, acetate NSC 152533 |
| InChI Formula | InChI=1S/C32H50O4/c1-19(2)21-11-16-32(27(34)35)18-17-30(7)22(26(21)32)9-10-24-29(6)14-13-25(36-20(3)33)28(4,5)23(29)12-15-31(24,30)8/h21-26H,1,9-18H2,2-8H3,(H,34,35)/t21-,22+,23-,24+,25-,26+,29-,30+,31+,32-/m0/s1 |
| SMILES | CC(=O)OC1CC[C@@]2(C)[C@@H](CC[C@]3(C)[C@@H]2CC[C@@H]4[C@H]5[C@@H](CC[C@@]5(CC[C@@]34C)C(=O)O)C(=C)C)C1(C)C |
| IUPAC Name | (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-acetyloxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid |
| Molecular Weight (g/mol) | 498.74 |