missing translation for 'onlineSavingsMsg'
Learn More

3-Nonen-2-one, 96%

Product Code. 11429386
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11429386

Brand: Thermo Scientific Alfa Aesar A19084.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14309-57-0 | C9H16O | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14309-57-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00010241
InChI Key HDKLIZDXVUCLHQ-BQYQJAHWSA-N
Synonym 3-nonen-2-one, trans-3-nonen-2-one, e-non-3-en-2-one, e-3-nonen-2-one, 3-nonen-2-one, 3e, 3e-3-nonen-2-one, 3e-non-3-en-2-one, unii-fr0k39of16, 3e-nonen-2-one, 3-nonen-2-one fhfi
PubChem CID 5317045
SMILES CCCCC\C=C\C(C)=O

Specifications

Density 0.848
Boiling Point 85°C (12 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.449
Quantity 50 g
Beilstein 1743558
IUPAC Name (E)-non-3-en-2-one
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 3-Nonen-2-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.