missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrophthalic-d3 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Specifications
Specifications
| Chemical Name or Material | 3-Nitrophthalic Acid-D3 |
| CAS | 1386387-74-1 |
| Molecular Formula | C8 D3 H2 N O6 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5-d3-dicarboxylic acid, 6-nitro- (ACI), 6-Nitro-1,2-benzene-3,4,5-d3-dicarboxylic acid (ACI), 3-Nitrophthalic-d3 Acid, 3-Nitrophthalic-4,5,6-d3 Acid, 3-Nitrophthalic Acid-D3 |
| InChI Formula | InChI=1 S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3 H,(H,10,11)(H,12,13)/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c(C(=O)O)c(c1[2 H])[N+](=O)[O-] |
| IUPAC Name | 3,4,5-trideuterio-6-nitrophthalic acid |
| Molecular Weight (g/mol) | 214.1469 |
| Formula Weight | 214.0305 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction