Learn More
3-Nitrobenzoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Nitrobenzoyl Chloride |
| CAS | 121-90-4 |
| Molecular Formula | C7H4ClNO3 |
| Quantity | 1 mg |
| Synonym | m-Nitrobenzoyl chloride |
| InChI Formula | InChI=1S/C7H4ClNO3/c8-7(10)5-2-1-3-6(4-5)9(11)12/h1-4H |
| SMILES | [O-][N+](=O)c1cccc(c1)C(=O)Cl |
| IUPAC Name | 3-nitrobenzoyl chloride |
| Molecular Weight (g/mol) | 185.56 |
| Formula Weight | 184.988 |