missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzoic-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | m-Nitrobenzoic-d4 Acid |
| CAS | 78399-78-7 |
| Molecular Formula | C7 D4 H N O4 |
| Quantity | 0.25 g |
| Synonym | Benzoic-2,3,4,6-d4 acid, 5-nitro- (9 CI), 3-Nitro-2,4,5,6-tetradeuteriobenzoic acid, 5-Nitrobenzoic-2,3,4,6-d4 acid, 3-Nitrobenzoic-2,4,5,6-d4 acid, m-Nitrobenzoic acid-d4, 3-Nitrobenzoic Acid-D4, 3-Nitrobenzoic-d4 Acid, m-Nitrobenzoic-d4 Acid |
| InChI Formula | InChI=1 S/C7H5NO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(c1[2 H])[N+](=O)[O-] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-nitrobenzoic acid |
| Molecular Weight (g/mol) | 171.144 |
| Formula Weight | 171.047 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction