missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-alpha-toluenesulfonyl chloride, 95%

Product Code. 11480947
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11480947

Brand: Thermo Scientific Alfa Aesar B23333.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58032-84-1 | C7H6ClNO4S | 235.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58032-84-1
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.64
MDL Number MFCD01859935
InChI Key IUYFKLDRPZLAQQ-UHFFFAOYSA-N
Synonym 3-nitrophenyl methanesulfonyl chloride, 3-nitrobenzylsulfonyl chloride, 3-nitrophenylmethanesulfonylchloride, benzenemethanesulfonyl chloride, 3-nitro, 3-nitro-alpha-toluenesulfonyl chloride, chloro 3-nitrophenyl methyl sulfone, acmc-1b04d, 3-nitro-phenylmethane-sulfonyl chloride, 3-nitrophenyl methanesulphonyl chloride, 3-nitrobenzenemethanesulfonyl chloride
PubChem CID 13196775
IUPAC Name (3-nitrophenyl)methanesulfonyl chloride
SMILES [O-][N+](=O)C1=CC=CC(CS(Cl)(=O)=O)=C1

Specifications

Melting Point 95°C to 102°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 235.64
Percent Purity 95%
Chemical Name or Material 3-Nitro-alpha-toluenesulfonyl chloride
compliance-icons
Product Identifier
  • 3-Nitrophenylmethanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 3-Nitrophenylmethanesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.