missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylindole-d8,NH, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-1 H-Indole-d8 |
| CAS | 697807-03-7 |
| Molecular Formula | C9 2H8 H N |
| Quantity | 0.01 g |
| Synonym | 1 H-Indole-2,4,5,6,7-d5, 3-(methyl-d3)-, 3-(Methyl-d3)-1 H-indole-2,4,5,6,7-d5, 3-Methyl-1 H-indole-d8, 3-Methylindole-D8, Scatole-d8, Skatol-d8, β-Methylindole-d8 |
| InChI Formula | InChI=1 S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,4,5,6,7-pentadeuterio-3-(trideuteriomethyl)-1 H-indole |
| Molecular Weight (g/mol) | 139.22 |
| Formula Weight | 139.1237 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction