Learn More
3-Methylbutyric-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 3-Methylbutanoic-2,2-D2 Acid |
| CAS | 95927-02-9 |
| Molecular Formula | C5H8D2O2 |
| Quantity | 0.5 g |
| Synonym | Butanoic-2,2-d2 acid, 3-methyl- (9 CI), 3-Methylbutyric-2,2-d2 acid, Isovaleric acid-2,2-d2, 3-Methylbutanoic-2,2-d2 acid, 3-Methylbutanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)/i3D2 |
| SMILES | CC(C([2 H])([2 H])C(O)=O)C |
| IUPAC Name | 3-methylbutanoic-2,2-d2 acid |
| Molecular Weight (g/mol) | 104.15 |
| Formula Weight | 104.08 g/mol |
| Show More |