Learn More
3-Methylamino-1-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methylamino-1-propanol |
| CAS | 42055-15-2 |
| Molecular Formula | C4H11NO |
| Quantity | 25 g |
| Synonym | (3-Hydroxypropyl)methylamine, 3-(Methylamino)propanol, 3-(N-Methylamino)-1-propanol, 3-(N-Methylamino)propanol, AMP-P 5, N-(3-Hydroxypropyl)-N-methylamine, N-(3-Hydroxypropyl)methylamine, N-Methyl-3-aminopropanol, N-Methylpropanolamine, gamma-(Methylamino)propanol |
| InChI Formula | InChI=1S/C4H11NO/c1-5-3-2-4-6/h5-6H,2-4H2,1H3 |
| SMILES | CNCCCO |
| IUPAC Name | 3-(methylamino)propan-1-ol |
| Molecular Weight (g/mol) | 89.14 |
| Formula Weight | 89.0841 |