Learn More
3-Methyl-5-phenylisoxazole, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-5-phenylisoxazole |
| CAS | 1008-75-9 |
| Molecular Formula | C10H9NO |
| Quantity | 50 mg |
| Synonym | 3-Methyl-5-phenyl-1,2-oxazole, NSC 80670 |
| InChI Formula | InChI=1S/C10H9NO/c1-8-7-10(12-11-8)9-5-3-2-4-6-9/h2-7H,1H3 |
| SMILES | CC1=NOC(=C1)C2=CC=CC=C2 |
| IUPAC Name | 3-methyl-5-phenyl-1,2-oxazole |
| Molecular Weight (g/mol) | 159.18 |
| Formula Weight | 159.0684 |