missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-benzyl-piperazine-d4, TRC

Product Code. 30474678
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474678 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30474678 Supplier Toronto Research Chemicals Supplier No. TRCM292842500MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-1-benzyl-piperazine-d4
CAS 1794752-53-6
Molecular Formula C12H14D4N2
Quantity 500 mg
Synonym 3-Methyl-1-(phenylmethyl)-piperazine-d4, 1-Benzyl-3-methyl-piperazine-d4, 3-Methyl-1-(phenylmethyl)piperazine-d4
InChI Formula InChI=1S/C12H18N2/c1-11-9-14(8-7-13-11)10-12-5-3-2-4-6-12/h2-6,11,13H,7-10H2,1H3/i7D2,9D2
SMILES [2H]C1([2H])CN(Cc2ccccc2)C([2H])([2H])C(C)N1
IUPAC Name 1-benzyl-2,2,5,5-tetradeuterio-3-methylpiperazine
Molecular Weight (g/mol) 194.31
Formula Weight 194.1721
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.