missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxytoluene-alpha,alpha,alpha-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Methoxy-3-methylbenzene-D3 (3-methyl-D3) |
| CAS | 20369-34-0 |
| Molecular Formula | C8 D3 H7 O |
| Quantity | 1 g |
| Synonym | Benzene, 1-methoxy-3-(methyl-d3)- (9 CI), Anisole, m-methyl-d3- (7 CI), 1-Methoxy-3-(trideuteriomethyl)benzene, 3-(Methyl-d3)-1-methoxybenzene, 3-Methoxytoluene-α,α,α-d3, 1-Methoxy-3-(methyl-d3)benzene, 1-Methoxy-3-methylbenzene-d3, 1-Methoxy-3-methylbenzene-D3 (3-methyl-D3) |
| InChI Formula | InChI=1 S/C8H10O/c1-7-4-3-5-8(6-7)9-2/h3-6 H,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cccc(OC)c1 |
| IUPAC Name | 1-methoxy-3-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 125.1829 |
| Formula Weight | 125.092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction