missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropane-1-sulfonyl Fluoride, TRC

Product Code. 30478411
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478411 50 mg 50mg
30484264 10 mg 10mg
30480524 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478411 Supplier Toronto Research Chemicals Supplier No. TRCM22682850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1227250-19-2
Molecular Formula C4H9O3FS
Molecular Weight (g/mol) 156.18
IUPAC Name 3-methoxypropane-1-sulfonyl fluoride
SMILES COCCCS(=O)(=O)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H9FO3S/c1-8-3-2-4-9(5,6)7/h2-4H2,1H3
Formula Weight 156.0256
Chemical Name or Material 3-Methoxypropane-1-sulfonyl Fluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.