missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methoxyphenyl)hydrazine Hydrochloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (3-Methoxyphenyl)hydrazine Hydrochloride |
| CAS | 39232-91-2 |
| Molecular Formula | C7H10N2O . (HCl) |
| Quantity | 1 g |
| Synonym | (3-Methoxyphenyl)hydrazine Hydrochloride, (3-Methoxyphenyl)hydrazine Monohydrochloride, [3-(Methyloxy)phenyl]hydrazine Monohydrochloride, m-Methoxyphenylhydrazine Hydrochloride |
| InChI Formula | InChI=1S/C7H10N2O.ClH/c1-10-7-4-2-3-6(5-7)9-8;/h2-5,9H,8H2,1H3;1H |
| SMILES | COC1=CC=CC(=C1)NN.Cl |
| IUPAC Name | (3-methoxyphenyl)hydrazine hydrochloride |
| Molecular Weight (g/mol) | 174.63 |
| Formula Weight | 174.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction