Learn More
3-Methoxybenzyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methoxybenzyl Chloride |
| CAS | 824-98-6 |
| Molecular Formula | C8 H9 Cl O |
| Quantity | 5 g |
| Synonym | 1-(Chloromethyl)-3-methoxybenzene |
| InChI Formula | InChI=1S/C8H9ClO/c1-10-8-4-2-3-7(5-8)6-9/h2-5H,6H2,1H3 |
| SMILES | COc1cccc(CCl)c1 |
| IUPAC Name | 1-(chloromethyl)-3-methoxybenzene |
| Molecular Weight (g/mol) | 156.61 |
| Formula Weight | 156.0342 |