Learn More
3-Methoxybenzyl Chloride, TRC
Code produit 30465340
Change view
Click to view available options
Quantity:
1 g
Conditionnement:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
30465340 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30465340 Fournisseur Toronto Research Chemicals Code fournisseur TRCM2611601G
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 3-Methoxybenzyl Chloride
CAS 824-98-6
Molecular Formula C8 H9 Cl O
Quantity 1 g
Synonym 1-(Chloromethyl)-3-methoxybenzene
InChI Formula InChI=1S/C8H9ClO/c1-10-8-4-2-3-7(5-8)6-9/h2-5H,6H2,1H3
SMILES COc1cccc(CCl)c1
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
Molecular Weight (g/mol) 156.61
Formula Weight 156.0342