Learn More
3-Methoxybenzyl alcohol, 98%
Product Code. 10726341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
Product Code. Quantity Packaging unitSize
10726341 25 mL Glass bottle 25mL
1 options
Product Code. 10726341
Brand: Thermo Scientific Acros 125770250
Request Bulk or Custom Format
CAS: 6971-51-3 | C8H10O2 | 138.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 6971-51-3
CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.1100g/mL
Boiling Point 250.0°C (723.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H10O2
Linear Formula CH3OC6H4CH2OH
Refractive Index 1.5425 to 1.5445
MDL Number MFCD00004637
Quantity 25 mL
Beilstein 06, 896
Specific Gravity 1.11
Synonym 3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol
Solubility Information Solubility in water: immiscible
InChI Key IIGNZLVHOZEOPV-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1)CO
IUPAC Name (3-methoxyphenyl)methanol
Molecular Weight (g/mol) 138.17
PubChem CID 81437
ChEBI CHEBI:1580
Formula Weight 138.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzyl alcohol, 98%