Learn More
3-Methoxybenzenemethanol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methoxybenzenemethanol |
| CAS | 6971-51-3 |
| Molecular Formula | C8H10O2 |
| Quantity | 5 g |
| Synonym | (3-Methoxyphenyl)methanol, 1-(Hydroxymethyl)-3-methoxybenzene, 3-Methoxybenzyl alcohol, [3-(Methyloxy)phenyl]methanol, m-Anisalcohol, m-Methoxybenzyl alcohol |
| InChI Formula | InChI=1S/C8H10O2/c1-10-8-4-2-3-7(5-8)6-9/h2-5,9H,6H2,1H3 |
| SMILES | COc1cccc(CO)c1 |
| IUPAC Name | (3-methoxyphenyl)methanol |
| Molecular Weight (g/mol) | 138.16 |
| Formula Weight | 138.0681 |