missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptophenylacetic Acid, 90%, TRC

Product Code. 30481324
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481324 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481324 Supplier Toronto Research Chemicals Supplier No. TRCM254500500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Mercaptophenylacetic Acid
CAS 63271-86-3
Molecular Formula C8H8O2S
Quantity 500 mg
Synonym 3-Mercapto-benzeneacetic Acid
InChI Formula InChI=1S/C8H8O2S/c9-8(10)5-6-2-1-3-7(11)4-6/h1-4,11H,5H2,(H,9,10)
SMILES OC(=O)Cc1cccc(S)c1
IUPAC Name 2-(3-sulfanylphenyl)acetic acid
Molecular Weight (g/mol) 168.2
Formula Weight 168.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.