Learn More
3-Keto-N-aminoethyl-N’-aminocaproyldihydrocinnamoyl Cyclopamine, TRC
Product Code. 30011548
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011548 5 mg 5mg
30010637 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011548 Supplier Toronto Research Chemicals Supplier No. TRCK1710005MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 306387-90-6
Molecular Formula C44 H63 N3 O4
Molecular Weight (g/mol) 697.99
IUPAC Name (3'R,3'aS,6'S,6aS,6bS,9R,11aS,11bR)-4'-[3-[4-[6-(2-aminoethylamino)hexanoyl]phenyl]propanoyl]-3',6',10,11b-tetramethylspiro[1,2,4,6,6a,6b,7,8,11,11a-decahydrobenzo[a]fluorene-9,2'-3,3a,5,6,7,7a-hexahydrofuro[3,2-b]pyridine]-3-one
SMILES C[C@H]1CC2O[C@]3(CC[C@H]4[C@@H]5CC=C6CC(=O)CC[C@]6(C)[C@H]5CC4=C3C)[C@H](C)[C@@H]2N(C1)C(=O)CCc7ccc(cc7)C(=O)CCCCCNCCN
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C44H63N3O4/c1-28-24-40-42(47(27-28)41(50)16-11-31-9-12-32(13-10-31)39(49)8-6-5-7-22-46-23-21-45)30(3)44(51-40)20-18-35-36-15-14-33-25-34(48)17-19-43(33,4)38(36)26-37(35)29(44)2/h9-10,12-14,28,30,35-36,38,40,42,46H,5-8,11,15-27,45H2,1-4H3/t28-,30+,35-,36-,38-,40?,42-,43-,44-/m0/s1
Formula Weight 697.4819
Chemical Name or Material 3-Keto-N-aminoethyl-N'-aminocaproyldihydrocinnamoyl Cyclopamine