Learn More
3-Iodo-2-propynyl N-Butylcarbamate-d9, TRC
Product Code. 30408743
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408743 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30408743 Supplier Toronto Research Chemicals Supplier No. TRCI7189525MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Iodo-2-propynyl N-Butylcarbamate-d9
CAS 1246815-08-6
Molecular Formula C8H3D9INO2
Quantity 5 mg
Synonym N-(Butylcarbamic Acid-d9) 3-Iodo-2-propyn-1-yl Ester, 3-Iodopropargyl n-(Butylcarbamate-d9), Acticide IPW 50-d9, Biodocarb-d9, Coatcide 123-d9, Dekaben LMB-d9, Ecobois-d9
InChI Formula InChI=1S/C8H12INO2/c1-2-3-6-10-8(11)12-7-4-5-9/h2-3,6-7H2,1H3,(H,10,11)
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC(=O)OCC#CI
Molecular Weight (g/mol) 290.15