missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyruvic Acid Lithium Salt (>80%), TRC

Product Code. 30406582
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406582 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406582 Supplier Toronto Research Chemicals Supplier No. TRCH95320050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxypyruvic Acid Lithium Salt
CAS 209728-15-4
Molecular Formula C3H3LiO4.H2O
Quantity 50 mg
Synonym 3-Hydroxy-2-oxopropanoic Acid Lithium Salt (1:1), Lithium 3-Hydroxypyruvate
InChI Formula InChI=1S/C3H4O4.Li/c4-1-2(5)3(6)7;/h4H,1H2,(H,6,7);/q;+1/p-1
SMILES O=C(C(O[Li])=O)CO.O
Molecular Weight (g/mol) 109.991802
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.