missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxybenzoic-2,4,5,6-d4 acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxybenzoic Acid-2,4,5,6-D4 |
| Molecular Formula | C7 D4 H2 O3 |
| Quantity | 0.25 g |
| Synonym | 5-Hydroxybenzoic-2,3,4,6-d4 acid), 3-Hydroxybenzoic-2,4,5,6-d4 acid, m-Salicylic acid-d4, 3-Hydroxybenzoic Acid-2,4,5,6-D4 |
| InChI Formula | InChI=1 S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8 H,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(O)c([2 H])c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 142.1454 |
| Formula Weight | 142.0568 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction