Learn More
3-Hydroxy Midostaurin, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy Midostaurin |
| CAS | 179237-49-1 |
| Molecular Formula | C35 H30 N4 O5 |
| Quantity | 2.5 mg |
| Synonym | N-(2,3,10,11,12,13-Hexahydro-3-hydroxy-10-methoxy-9-methyl-1-oxo-9,13-epoxy-1H,9H-diindolo[1,2,3-gh:3',2',1'-lm]pyrrolo[3,4-j][1,7]benzodiazonin-11-yl)-N-methylbenzamide, CGP 52421 |
| InChI Formula | InChI=1S/C35H30N4O5/c1-35-31(43-3)23(37(2)34(42)18-11-5-4-6-12-18)17-24(44-35)38-21-15-9-7-13-19(21)25-27-28(33(41)36-32(27)40)26-20-14-8-10-16-22(20)39(35)30(26)29(25)38/h4-16,23-24,31,33,41H,17H2,1-3H3,(H,36,40)/t23-,24?,31-,33?,35+/m1/s1 |
| SMILES | CO[C@@H]1[C@@H](CC2O[C@]1(C)n3c4ccccc4c5c6C(O)NC(=O)c6c7c8ccccc8n2c7c35)N(C)C(=O)c9ccccc9 |
| Molecular Weight (g/mol) | 586.64 |
| Formula Weight | 586.2216 |