Learn More
3-Hydroxy-2,2-dimethylpropanal, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-2,2-dimethylpropanal |
| CAS | 597-31-9 |
| Molecular Formula | C5H10O2 |
| Quantity | 5 g |
| Synonym | 2,2-Dimethyl-3-hydroxypropanal, 2,2-Dimethyl-3-hydroxypropionaldehyde, 2-(Hydroxymethyl)-2-methylpropanal, 2-(Hydroxymethyl)isobutyraldehyde, 3-Hydroxy-2,2-dimethylpropionaldehyde, 3-Hydroxypivalaldehyde, Hydroxypivalaldehyde, Hydroxypivaldehyde, Pentaaldol, Pentaldol, α,α-Dimethyl-β-hydroxypropionaldehyde |
| InChI Formula | InChI=1S/C5H10O2/c1-5(2,3-6)4-7/h3,7H,4H2,1-2H3 |
| SMILES | CC(C)(CO)C=O |
| IUPAC Name | 3-hydroxy-2,2-dimethylpropanal |
| Molecular Weight (g/mol) | 102.13 |
| Formula Weight | 102.0681 |