Learn More
3-Hydroxy-1-methyl-pyridinium Bromide, TRC
Product Code. 30401812
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30401812 Supplier Toronto Research Chemicals Supplier No. TRCH9480081G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxy-1-methylpyridin-1-ium bromide
CAS 31034-86-3
Molecular Formula C6H8NO.Br
Quantity 1 g
Synonym 3-Hydroxy-1-methylpyridinium Bromide, Pyridinium, 3-hydroxy-1-methyl-, bromide (1:1), 3-Hydroxy-1-methylpyridinium bromide (6CI), Pyridinium, 3-hydroxy-1-methyl-, bromide (8CI,9CI), 3-Hydroxy-N-methylpyridinium bromide, N-Methyl-3-hydroxypyridinium bromide, Ro 1-5237
InChI Formula InChI=1S/C6H7NO.BrH/c1-7-4-2-3-6(8)5-7;/h2-5H,1H3;1H
SMILES [Br-].C[n+]1cccc(O)c1
IUPAC Name 1-methylpyridin-1-ium-3-ol;bromide
Molecular Weight (g/mol) 190.04
Formula Weight 188.979