Learn More
3-Hydroxy-1-methyl-pyridinium Bromide, TRC
Product Code. 30396468
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396468 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30396468 Supplier Toronto Research Chemicals Supplier No. TRCH948008250MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxy-1-methylpyridin-1-ium bromide
CAS 31034-86-3
Molecular Formula C6H8NO.Br
Quantity 250 mg
Synonym 3-Hydroxy-1-methylpyridinium Bromide, Pyridinium, 3-hydroxy-1-methyl-, bromide (1:1), 3-Hydroxy-1-methylpyridinium bromide (6CI), Pyridinium, 3-hydroxy-1-methyl-, bromide (8CI,9CI), 3-Hydroxy-N-methylpyridinium bromide, N-Methyl-3-hydroxypyridinium bromide, Ro 1-5237
InChI Formula InChI=1S/C6H7NO.BrH/c1-7-4-2-3-6(8)5-7;/h2-5H,1H3;1H
SMILES [Br-].C[n+]1cccc(O)c1
IUPAC Name 1-methylpyridin-1-ium-3-ol;bromide
Molecular Weight (g/mol) 190.04
Formula Weight 188.979