Learn More
3-Formyl-4-nitrobenzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Formyl-4-nitrobenzonitrile |
| CAS | 90178-82-8 |
| Molecular Formula | C8H4N2O3 |
| Quantity | 100 mg |
| Synonym | 5-Cyano-2-nitrobenzaldehyde |
| InChI Formula | InChI=1S/C8H4N2O3/c9-4-6-1-2-8(10(12)13)7(3-6)5-11/h1-3,5H |
| SMILES | [O-][N+](=O)c1ccc(cc1C=O)C#N |
| IUPAC Name | 3-formyl-4-nitrobenzonitrile |
| Molecular Weight (g/mol) | 176.13 |
| Formula Weight | 176.0222 |