Learn More
3-Fluorothiophenol, 98%
Product Code. 11476326
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476326 1 g 1g
11486326 5 g 5g
11496326 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11476326 Supplier Thermo Scientific Alfa Aesar Supplier No. B23114.03
 Restricted Item
In Stock
Add to basket
CAS: 2557-77-9 | C6H5FS | 128.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2557-77-9
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.16
MDL Number MFCD00040227
InChI Key ZDEUGINAVLMAET-UHFFFAOYSA-N
Synonym 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol
PubChem CID 75710
IUPAC Name 3-fluorobenzenethiol
SMILES FC1=CC=CC(S)=C1
Specifications
Density 1.203
Boiling Point 156°C
Flash Point 58°C (136°F)
Odor Stench
Refractive Index 1.554
Quantity 1 g
UN Number UN3071
Beilstein 2039302
Sensitivity Air Sensitive
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3-Fluorothiophenol