Learn More
3-Fluorophenyl acetate, 98%, Thermo Scientific™
Product Code. 11383087
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383087 25 g 25g
11373087 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11383087 Supplier Thermo Scientific Alfa Aesar Supplier No. L13465.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 701-83-7
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142578
InChI Key IAWZWMGUTKRLQB-UHFFFAOYSA-N
Synonym 1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
PubChem CID 136542
IUPAC Name (3-fluorophenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)F
Specifications
Density 1.15
Boiling Point 77°C to 78°C (13 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.478
Quantity 25 g
Beilstein 2555261
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluorophenyl acetate