Learn More
3-Fluorobenzonitrile, 98%
Product Code. 11485397
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11485397
Brand: Thermo Scientific Alfa Aesar B23426.14
Request Bulk or Custom Format
CAS: 403-54-3 | C7H4FN | 121.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N
Specifications
Melting Point -16°C
Density 1.14
Boiling Point 183°C
Flash Point 67°C (152°F)
Odor Almond-like
Refractive Index 1.5055
Quantity 25 g
UN Number UN3276
Beilstein 1858941
Formula Weight 121.11
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzonitrile