Learn More
3-Fluoro-4-iodobenzonitrile, 97%, Thermo Scientific™
Product Code. 15421298
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15421298
Brand: Thermo Scientific Alfa Aesar H55141.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 887266-99-1
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.011
MDL Number MFCD07782076
InChI Key NPKQMCCUPXZXFI-UHFFFAOYSA-N
PubChem CID 24721521
IUPAC Name 3-fluoro-4-iodobenzonitrile
SMILES C1=CC(=C(C=C1C#N)F)I
Specifications
Melting Point 113°C to 117°C
Quantity 250 mg
Sensitivity Light sensitive
Synonym benzonitrile, 3-fluoro-4-iodo, benzonitrile,3-fluoro-4-iodo, 4-cyano-2-fluoroiodobenzene, acmc-20aiba, pubchem4781, 5-fluoro-4-iodobenzonitrile, 3-fluoro-4-iodo benzonitrile, 3-fluoro-4-iodo-benzonitrile, 3-fluoro-4-iodobenzonitrile3, ksc661e6h
Formula Weight 247.01
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-iodobenzonitrile