Learn More
3-Ethynylaniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 54060-30-9 |
| Molecular Formula | C8H7N |
| MDL Number | MFCD00014779 |
| Quantity | 10 g |
| SMILES | C1=C(C=CC=C1C#C)N |
| Molecular Weight (g/mol) | 117.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Ethynylaniline |