Learn More
3-Ethyl-2-methylpentane, TRC
Product Code. 30374699
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374699 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30374699 Supplier Toronto Research Chemicals Supplier No. TRCE93924350MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Ethyl-2-methylpentane
CAS 609-26-7
Molecular Formula C8 H18
Quantity 50 mg
InChI Formula InChI=1S/C8H18/c1-5-8(6-2)7(3)4/h7-8H,5-6H2,1-4H3
SMILES CCC(CC)C(C)C
IUPAC Name 3-ethyl-2-methylpentane
Molecular Weight (g/mol) 114.228
Formula Weight 114.141