missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-imine Meloxicam, TRC

Product Code. 30370854
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370854 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370854 Supplier Toronto Research Chemicals Supplier No. TRCE919950250MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(2Z)-3-Ethyl-5-methylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 1331636-17-9
Molecular Formula C16 H17 N3 O4 S2
Quantity 250 mg
Synonym N-[(2Z)-3-Ethyl-5-methylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Meloxicam Imp. D (EP), Meloxicam Imp. D (EP), N-[(2Z)-3-Ethyl-5-methylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Meloxicam Impurity D, 3-Ethyl-2-imine Meloxicam
InChI Formula InChI=1S/C16H17N3O4S2/c1-4-19-9-10(2)24-16(19)17-15(21)13-14(20)11-7-5-6-8-12(11)25(22,23)18(13)3/h5-9,20H,4H2,1-3H3/b17-16-
SMILES CCN1C=C(C)S/C/1=N\C(=O)C2=C(O)c3ccccc3S(=O)(=O)N2C
IUPAC Name (NZ)-N-(3-ethyl-5-methyl-1,3-thiazol-2-ylidene)-4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 379.45
Formula Weight 379.066
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.