missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10505300 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10505300

Brand: Thermo Scientific Acros 433190010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 90555-66-1
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.99
InChI Key CHCWUTJYLUBETR-UHFFFAOYSA-N
Synonym 3-ethoxyphenyl boronic acid, 3-ethoxybenzeneboronic acid, 3-ethoxyphenylboronicacid, 3-ethoxyphenyl boranediol, m-ethoxyphenylboronic acid, boronic acid, 3-ethoxyphenyl, pubchem1835, acmc-209r6q, 3-ethoxy-phenylboronic acid
PubChem CID 2773920
IUPAC Name (3-ethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)OCC)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151.5°C to 152.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 165.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.